MilliporeSigma
  • Characterization of PLGA based composite nerve guidance conduits: effect of F127 content on modulus over time in simulated physiological conditions.
Characterization of PLGA based composite nerve guidance conduits: effect of F127 content on modulus over time in simulated physiological conditions.
Journal of the mechanical behavior of biomedical materials (2012-10-03)
S Kehoe, X F Zhang, L Lewis, H O'Shea, D Boyd
ABSTRACT
PLGA/pluronic F127 based nerve guidance conduits (NGCs) for peripheral nerve regeneration offer excellent potential for clinical use. To date, little emphasis has been directed towards the effect of pluronic F127 on their subsequent mechanical properties as a function of degradation time or the physiological environment. This report was designed to redress the balance. This study synthesised 5 groups of 20wt% PLGA NGCs with varied additions of pluronic F127 (range 0-5wt%) to obtain Young's Moduli (E) in the range of 7-107MPa, depending on degradation conditions and pluronic F127 content.
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol-ran-propylene glycol) monobutyl ether, average Mn ~3,900
Sigma-Aldrich
Tetraglycol, BioXtra