MilliporeSigma
  • Cyclic tetrapeptides with thioacetate tails or intramolecular disulfide bridge as potent inhibitors of histone deacetylases.
Cyclic tetrapeptides with thioacetate tails or intramolecular disulfide bridge as potent inhibitors of histone deacetylases.
Bioorganic & medicinal chemistry letters (2012-10-02)
Md Ashraful Hoque, Toru Arai, Norikazu Nishino, Hyun-Jung Kim, Akihiro Ito, Minoru Yoshida
ABSTRACT
Two thioacetate tails were introduced to the chlamydocin- and CHAP31-related cyclic tetrapeptides. An intramolecular disulfide bridge could be formed in the CHAP31-related cyclic peptides. Both the thioacetate-tailed and disulfide-bridged peptides were potent histone deacetylase inhibitors in the presence of sulfhydryl compound. Potent p21 promoter inducing activity was also observed in vivo.
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Thioacetic acid, 96%
Sigma-Aldrich
Potassium thioacetate, 98%