MilliporeSigma
  • Anoxia-induced release of colloid- and nanoparticle-bound phosphorus in grassland soils.
Anoxia-induced release of colloid- and nanoparticle-bound phosphorus in grassland soils.
Environmental science & technology (2012-09-29)
R Henderson, N Kabengi, N Mantripragada, M Cabrera, S Hassan, A Thompson
ABSTRACT
Particle-facilitated transport is a key mechanism of phosphorus (P) loss in agroecosystems. We assessed contributions of colloid- and nanoparticle-bound P (nPP; 1-415 nm) to total P released from grassland soils receiving biannual poultry litter applications since 1995. In laboratory incubations, soils were subjected to 7 days of anoxic conditions or equilibrated at pH 6 and 8 under oxic conditions and then the extract was size fractionated by differential centrifugation/ultrafiltration for analysis of P, Al, Fe, Si, Ti, and Ca. Selected samples were characterized by transmission electron microscopy-energy dispersive spectroscopy (TEM-EDS) and field flow fractionation (FFF-ICP-MS). Particles released were present as nanoaggregates with a mean diameter of 200-250 nm, composed of ~50-nm aluminosilicate flakes studded with Fe and Ti-rich clusters (<10 nm) that contained most of the P detected by EDS. Anoxic incubation of stimulated nPP release with seasonally saturated soils released more nPP and Fe(2+)(aq) than well-drained soils; whereas, nonreductive particle dispersion, accomplished by raising the pH, yielded no increase in nPP release. This suggests Fe acts as a cementing agent, binding to the bulk soil P-bearing colloids that can be released during reducing conditions. Furthermore, it suggests prior periodic exposure to anoxic conditions increases susceptibility to redox-induced P mobilization.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets