MilliporeSigma
  • Intrinsic single-band upconversion emission in colloidal Yb/Er(Tm):Na3Zr(Hf)F7 nanocrystals.
Intrinsic single-band upconversion emission in colloidal Yb/Er(Tm):Na3Zr(Hf)F7 nanocrystals.
Chemical communications (Cambridge, England) (2012-09-27)
Daqin Chen, Lei Lei, Rui Zhang, Anping Yang, Ju Xu, Yuansheng Wang
ABSTRACT
Novel Yb/Er(Tm):Na(3)MF(7) (M = Zr, Hf) nanocrystals with intrinsic single-band upconversion emission, in contrast to the routine lanthanide-doped fluoride nanocrystals which show typical multi-band upconversion emissions, are reported for the first time. Specifically, the red upconversion intensity of the Yb/Er:Na(3)ZrF(7) nanocrystals is about 5 times as high as that of the hexagonal Yb/Er:NaYF(4) ones with a similar crystal size.
Sigma-Aldrich
Hafnium, turnings, crystal bar, 99.7% trace metals basis