MilliporeSigma
  • Superacid-promoted ionization of alkanes without carbonium ion formation: a density functional theory study.
Superacid-promoted ionization of alkanes without carbonium ion formation: a density functional theory study.
The journal of physical chemistry. A (2012-09-25)
Peter Dinér
ABSTRACT
The carbonium ion has been suggested to be the intermediate in superacid-promoted reactions (SbF(5)-HF) such as hydrogen-deuterium exchange and in the electrophilic C-H cleavage into hydrogen and the carbenium ion. In this study, the superacid-promoted C-H cleavage into hydrogen and the carbenium ion was studied using density functional theory (B3LYP and M062X) and ab initio methods (MP2 and CCSD). The calculations suggest that the superacid-promoted C-H cleavage proceeds via a concerted transition state leading to hydrogen (H(2)) and the carbenium ion without the formation of the elusive carbonium ion. The reactivity for the superacid-promoted C-H cleavage decreases upon going from isobutane (tertiary) > propane (secondary) > isobutane (primary) > propane (primary) > ethane > methane.
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis