MilliporeSigma
  • New filtering antimicrobial nonwovens with various carriers for biocides as respiratory protective materials against bioaerosol.
New filtering antimicrobial nonwovens with various carriers for biocides as respiratory protective materials against bioaerosol.
International journal of occupational safety and ergonomics : JOSE (2012-09-22)
Katarzyna Majchrzycka, Beata Gutarowska, Agnieszka Brochocka, Bogumił Brycki
ABSTRACT
This study evaluated the bioactivity of polypropylene melt-blown filtering nonwovens used in respiratory protective devices (RPD) with a biocidal agent (alkylammonium microbiocides) on 2 mineral carriers. Two types of carriers were tested: a bentonite, with an aluminosilicate base, and a perlite, volcanic glass. High biostatic and biocidal effects of modified nonwovens with biocides were tested against Escherichia coli (E. coli) and Staphylococcus aureus (S. aureus) bacteria. Nonwovens modified with a biocide on a bentonite carrier showed an opposite reaction to a biocide on a perlite. The research also showed that 10% concentration of a biocidal agent on a perlite carrier was sufficient to inhibit the growth of bacteria (100% reduction) placed in the structure of a filtering material during normal use of RPD. A comparison of the biological activity of 2 filtering materials, each containing 10% of a perlite and produced in a laboratory and industrial conditions, showed no statistically significant differences.
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
Sigma-Aldrich
Bentonite