MilliporeSigma
  • Comparison of changes in facial skin temperature caused by ethyl chloride spraying, ice block rubbing and cold gel packing in healthy subjects.
Comparison of changes in facial skin temperature caused by ethyl chloride spraying, ice block rubbing and cold gel packing in healthy subjects.
Journal of oral rehabilitation (2012-09-22)
Y-G Im, H-J Park, H-Y Chae, B-G Kim, H-S Lim, J-I Park, J-H Kim
ABSTRACT
The aim of this study was to compare the effects of three cryotherapeutic modalities (ethyl chloride spraying, ice block rubbing and cold gel packing) on facial skin temperature. Thirty healthy volunteers (15 men, 15 women; mean age, 29·4 ± 3·2 years) participated in this study. Each of the three modalities was randomly applied to the skin over the right masseter muscle. The skin surface temperature was recorded at baseline and every 5 min for 60 min after the application of one of the three cryotherapeutic modalities. Immediately after application, cold gel packing demonstrated the greatest reduction in surface temperature (10·6 °C), followed by ethyl chloride spraying (4·3 °C) and ice block rubbing (3·7 °C) (P < 0·001). During the 60-min post-application period, ethyl chloride spraying and ice block rubbing produced similar skin surface temperature changes. The skin surface remained coldest for the longest period of time after cold gel packing. The median time for recovery of the baseline temperature after application of the cold gel pack was about three to four times longer than that for the other modalities (P < 0·001). Ethyl chloride spraying and ice block rubbing resulted in less reduction and faster recovery of skin surface temperature than did cold gel packing. In conclusion, ethyl chloride spraying and ice block rubbing had a limited cooling effect on the facial skin tissue and could not reduce the skin surface temperature enough for local analgesia. Moreover, the cooling effect of cold gel packing was remarkable, but not sufficient for local analgesia.