MilliporeSigma
Time resolved polarization dependent single shot four wave mixing.
Physical chemistry chemical physics : PCCP (2012-09-20)
Andrey Shalit, Yehiam Prior
ABSTRACT
Single shot time resolved four wave mixing is a powerful tool for the acquisition of dynamic and spectroscopic data from molecules susceptible to bleaching or other photo-induced damage. We add polarization dependence to single shot methods, and demonstrate how magic angle measurements are made simpler by this methodology. We propose a new approach to single shot combined time/polarization measurements which can be generalized to other two dimensional combinations.
Product Description
Supelco
Dibromomethane, analytical standard
Sigma-Aldrich
Dibromomethane, 99%