MilliporeSigma
Glycine rescue of β-sheets from cis-proline.
Journal of the American Chemical Society (2012-09-20)
Madhurima Das, Gautam Basu
ABSTRACT
Proline is incompatible with ideal β-sheet geometry, and the incompatibility gets magnified when Pro assumes the cis peptidyl-prolyl conformation. We show that Gly appears with high propensity at pre-cisPro positions in β-sheets and rescues the β-sheet from severe distortions by assuming a right-handed polyproline conformation (β(PR)), effectively increasing the local β-sheet register by one residue. The united residue, Gly(β(PR))-cisPro, is evolutionarily conserved, functionally important, and dynamic in nature.
Sigma-Aldrich
Poly-L-proline, mol wt >30,000
Sigma-Aldrich
Poly-L-proline, mol wt 1,000-10,000