MilliporeSigma
  • Severe pollution of PCDD/Fs and dioxin-like PCBs in sediments from Lake Shihwa, Korea: tracking the source.
Severe pollution of PCDD/Fs and dioxin-like PCBs in sediments from Lake Shihwa, Korea: tracking the source.
Marine pollution bulletin (2012-09-18)
Hyo-Bang Moon, Minkyu Choi, Hee-Gu Choi, Kurunthachalam Kannan
ABSTRACT
Concentrations of polychlorinated dibenzo-p-dioxins, dibenzofurans (PCDD/Fs) and dioxin-like polychlorinated biphenyls (DL-PCBs) were determined in surface sediments from Lake Shihwa and from creeks that discharge into this artificial lake. The toxic equivalents (TEQs) in sediments ranged from 1.0 to 1770 pg/g dry weight, which were some of the highest values ever reported so far for coastal sediments on a global basis. The concentrations of PCDD/Fs and DL-PCBs decreased with increasing distance from the creeks to offshore regions of the lake. Based on a multivariate statistical analysis and congener profiles, it was found that high contamination by PCDD/Fs in creek sediments collected around Lake Shihwa was associated with releases from chlor-alkali processes. Significant correlations were found for total organic carbon content and the concentrations of highly chlorinated PCDD/F congeners. This is the first report to document chlor-alkali processes as a significant source of PCDD/F contamination in Korean coastal waters.
Product Description
Dibenzo[b,d]furan, BCR®, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dibenzofuran, 98%