MilliporeSigma
Electronic structure and magnetism of La4Ni3O8 from first principles.
Journal of physics. Condensed matter : an Institute of Physics journal (2012-09-13)
Ting Liu, Guoren Zhang, Xiaoli Zhang, Ting Jia, Zhi Zeng, H Q Lin
ABSTRACT
The magnetic and electronic properties of La(4)Ni(3)O(8) are investigated by performing the full-potential linearized augmented plane wave method. The C-type antiferromagnetic spin ordering is preferred and a molecular correlated insulating state with high-spin Ni ions is found. Our results have proved that this insulating state is caused by a correlation effect and the strong interlayer interaction. Such strong interlayer coupling results from the high-spin occupation of Ni ions.
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Nickel(II) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis