MilliporeSigma
  • Polydatin (PD) inhibits IgE-mediated passive cutaneous anaphylaxis in mice by stabilizing mast cells through modulating Ca²⁺ mobilization.
Polydatin (PD) inhibits IgE-mediated passive cutaneous anaphylaxis in mice by stabilizing mast cells through modulating Ca²⁺ mobilization.
Toxicology and applied pharmacology (2012-09-11)
Meichun Yuan, Jianjie Li, Jingzhang Lv, Xucheng Mo, Chengbin Yang, Xiangdong Chen, Zhigang Liu, Jie Liu
ABSTRACT
Mast cells play a key role in the pathogenesis of asthma and are a promising target for therapeutic intervention in asthma. This study investigated the effects of polydatin (PD), a resveratrol glucoside, on mast cell degranulation upon cross-linking of the high-affinity IgE receptors (FcεRI), as well as the anti-allergic activity of PD in vivo. Herein, we demonstrated that PD treatment for 30 min suppressed FcεRI-mediated mast cell degranulation in a dose-dependent manner. Concomitantly, PD significantly decreased FcεRI-mediated Ca²⁺ increase in mast cells. The suppressive effects of PD on FcεRI-mediated Ca²⁺ increase were largely inhibited by using LaCl₃ to block the Ca²⁺ release-activated Ca²⁺ channels (CRACs). Furthermore, PD significantly inhibited Ca²⁺ entry through CRACs evoked by thapsigargin (TG). Knocking down protein expression of Orai1, the pore-forming subunit of CRACs, significantly decreased PD suppression of FcεRI-induced intracellular Ca²⁺ influx and mast cell degranulation. In a mouse model of mast cell-dependent passive cutaneous anaphylaxis (PCA), in vivo PD administration suppressed mast cell degranulation and inhibited anaphylaxis. Taken together, our data indicate that PD stabilizes mast cells by suppressing FcεRI-induced Ca²⁺ mobilization mainly through inhibiting Ca²⁺ entry via CRACs, thus exerting a protective effect against PCA.
Sigma-Aldrich
Polydatin, ≥95% (HPLC)