MilliporeSigma
  • Enrichment of eggs in n-3 polyunsaturated fatty acids by feeding hens with different amount of linseed oil in diet.
Enrichment of eggs in n-3 polyunsaturated fatty acids by feeding hens with different amount of linseed oil in diet.
Food chemistry (2012-09-08)
Marinko Petrović, Milica Gačić, Veseljko Karačić, Zeljko Gottstein, Hrvoje Mazija, Helga Medić
ABSTRACT
The production of n-3 polyunsaturated fatty acid enriched eggs by addition of linseed oil to the laying hens' diet has been evaluated in terms of production parameters and n-6/n-3 ratio. A total of 150 18weeks old Lohmann Brown laying hens were housed in cages and fed with basal diet and four experimental diets containing 1%, 2%, 3% or 4% of linseed oil added to the basal diet. The effect of the altered level of linseed oil on hens laying performance, fatty acid content and composition and cholesterol content in egg yolk has been evaluated during 13weeks of experiment. Egg weight, yolk fat content, yolk weight, yolk percentage and shape index were not influenced by dietary treatment. The ratio between n-6 and n-3 polyunsaturated fatty acids in eggs decreased in first 5weeks and then remained stable until the end of the experiment for all experimental groups. Different contents of linseed oil in feed highly influenced the n-6/n-3 ratio (P<0.0001). Addition of linseed oil did not influence the cholesterol content in yolks (P=0.5200) while the only factor affecting the cholesterol content was the hens age (P<0.0001).
Sigma-Aldrich
Linseed Oil