MilliporeSigma
  • Inhibitors of reactive oxygen species accumulation delay and/or reduce the lethality of several antistaphylococcal agents.
Inhibitors of reactive oxygen species accumulation delay and/or reduce the lethality of several antistaphylococcal agents.
Antimicrobial agents and chemotherapy (2012-09-06)
Yuanli Liu, Xinghan Liu, Yilin Qu, Xiuhong Wang, Liping Li, Xilin Zhao
ABSTRACT
Perturbation of hydroxyl radical accumulation by subinhibitory concentrations of 2,2'-bipyridyl plus thiourea protects Escherichia coli from being killed by 3 lethal antimicrobial classes. Here, we show that 2,2'-bipyridyl plus thiourea delays and/or reduces antimicrobial killing of Staphylococcus aureus by daptomycin, moxifloxacin, and oxacillin. While the protective effect of 2,2'-bipyridyl plus thiourea varied among strains and compounds, the data support the hypothesis that hydroxyl radical enhances antimicrobial lethality.
Sigma-Aldrich
Oxacillin sodium salt, 815-950 μg/mg (Oxacillin)
Supelco
Oxacillin sodium salt monohydrate, VETRANAL®, analytical standard