MilliporeSigma
  • Spiro-fused pyrrolidine, piperidine, and oxindole scaffolds from lactams.
Spiro-fused pyrrolidine, piperidine, and oxindole scaffolds from lactams.
Organic letters (2012-09-06)
Christoph Hirschhäuser, Jeremy S Parker, Matthew W D Perry, Mairi F Haddow, Timothy Gallagher
ABSTRACT
Expedient routes to three classes of novel spiro-fused pyrrolidine, piperidine, and indoline heterocycle scaffolds are described. These three-dimensional frameworks, which conform to the "rule of three", are suitably protected to allow for site-selective manipulation and functionalization. Different modes of substrate control were explored, which allow for good to excellent levels of diastereoselectivity and dispense with the need for additional chiral reagents or catalysts. The concepts developed were applied in short, formal syntheses of (±)-coerulescine and (±)-horsfiline.
Sigma-Aldrich
Piperidine, puriss. p.a., ≥99.0% (GC/T)
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrrolidine, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
Please contact Customer Service for Availability
$41.10
100 mL
Please contact Customer Service for Availability
$57.60
500 mL
Please contact Customer Service for Availability
$151.00
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
Please contact Customer Service for Availability
$56.30
100 mL
Please contact Customer Service for Availability
$60.20
500 mL
Please contact Customer Service for Availability
$143.00
4 x 100 mL
Please contact Customer Service for Availability
$200.00
1 L
Please contact Customer Service for Availability
$235.00
2.5 L
Please contact Customer Service for Availability
$437.00
Sigma-Aldrich
Pyrrolidine, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
$74.30
1 kg
Please contact Customer Service for Availability
$119.00
8 kg
Please contact Customer Service for Availability
$595.00
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 mL
Please contact Customer Service for Availability
$27.30
100 mL
Please contact Customer Service for Availability
$48.00
500 mL
Please contact Customer Service for Availability
$169.00
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
200 mL
Please contact Customer Service for Availability
$140.00
450 mL
Please contact Customer Service for Availability
$246.00
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
$108.00
1 L
Please contact Customer Service for Availability
$586.00
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
$88.80
1 L
Please contact Customer Service for Availability
$379.00
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mL
Please contact Customer Service for Availability
$255.00
2 L
Please contact Customer Service for Availability
$751.00