MilliporeSigma
  • The SUMO protease SENP1 is required for cohesion maintenance and mitotic arrest following spindle poison treatment.
The SUMO protease SENP1 is required for cohesion maintenance and mitotic arrest following spindle poison treatment.
Biochemical and biophysical research communications (2012-09-05)
Saho Era, Takuya Abe, Hiroshi Arakawa, Shunsuke Kobayashi, Barnabas Szakal, Yusuke Yoshikawa, Akira Motegi, Shunichi Takeda, Dana Branzei
ABSTRACT
SUMO conjugation is a reversible posttranslational modification that regulates protein function. SENP1 is one of the six SUMO-specific proteases present in vertebrate cells and its altered expression is observed in several carcinomas. To characterize SENP1 role in genome integrity, we generated Senp1 knockout chicken DT40 cells. SENP1(-/-) cells show normal proliferation, but are sensitive to spindle poisons. This hypersensitivity correlates with increased sister chromatid separation, mitotic slippage, and apoptosis. To test whether the cohesion defect had a causal relationship with the observed mitotic events, we restored the cohesive status of sister chromatids by introducing the TOP2α(+/-) mutation, which leads to increased catenation, or by inhibiting Plk1 and Aurora B kinases that promote cohesin release from chromosomes during prolonged mitotic arrest. Although TOP2α is SUMOylated during mitosis, the TOP2α(+/-) mutation had no obvious effect. By contrast, inhibition of Plk1 or Aurora B rescued the hypersensitivity of SENP1(-/-) cells to colcemid. In conclusion, we identify SENP1 as a novel factor required for mitotic arrest and cohesion maintenance during prolonged mitotic arrest induced by spindle poisons.
Sigma-Aldrich
Demecolcine, ≥98% (HPLC)
Sigma-Aldrich
Demecolcine solution, 10 μg/mL in HBSS, ACF Qualified, BioXtra