MilliporeSigma
  • Paenibacillus catalpae sp. nov., isolated from the rhizosphere soil of Catalpa speciosa.
Paenibacillus catalpae sp. nov., isolated from the rhizosphere soil of Catalpa speciosa.
International journal of systematic and evolutionary microbiology (2012-09-04)
Jian Zhang, Zi-Ting Wang, Hui-Min Yu, Yuchao Ma
ABSTRACT
A bacterial strain, designated D75(T), was isolated from the rhizosphere soil of Catalpa speciosa. Phylogenetic analysis based on the complete 16S rRNA gene sequence revealed that strain D75(T) was a member of the genus Paenibacillus. High levels of 16S rRNA gene sequence similarity were found between strain D75(T) and Paenibacillus glycanilyticus DS-1(T) (99.2 %), Paenibacillus xinjiangensis B538(T) (97.5 %) and Paenibacillus castaneae Ch-32(T) (97.2 %). The chemotaxonomic properties of strain D75(T) were consistent with those of the genus Paenibacillus: the cell-wall peptidoglycan type was based on meso-diaminopimelic acid (A1γ), the predominant menaquinone was MK-7, and the major fatty acids were anteiso-C15 : 0, iso-C16 : 0 and C16 : 0. However, levels of DNA-DNA relatedness between strain D75(T) and P. glycanilyticus NBRC 16618(T), P. xinjiangensis DSM 16970(T) and P. castaneae DSM 19417(T) were 35, 20 and 18 %, respectively. On the basis of phenotypic and chemotaxonomic analyses, phylogenetic data and DNA-DNA relatedness values, strain D75(T) is considered to represent a novel species of the genus Paenibacillus, for which the name Paenibacillus catalpae sp. nov. is proposed. The type strain is D75(T) ( = DSM 24714(T) = CGMCC 1.10784(T)).
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥95.0% (NT)
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥98% (TLC)