MilliporeSigma
  • Isolation and characterization of a lipopeptide bioemulsifier produced by Pseudomonas nitroreducens TSB.MJ10 isolated from a mangrove ecosystem.
Isolation and characterization of a lipopeptide bioemulsifier produced by Pseudomonas nitroreducens TSB.MJ10 isolated from a mangrove ecosystem.
Bioresource technology (2012-09-04)
Trelita de Sousa, Saroj Bhosle
ABSTRACT
Pseudomonas nitroreducens TSB.MJ10 exhibiting growth and bioemulsifier production with 0.5% sodium benzoate as the sole carbon source was isolated from a mangrove ecosystem in the vicinity of a petroleum pump. The bioemulsifier is a lipopeptide that is stable over a pH range of 5-11 and a temperature range of 20-90°C and showed emulsifying activity in the presence of relatively high NaCl concentrations (up to 25%). The bioemulsifier formed stable emulsions with aliphatic (hexadecane, n-heptane, cyclohexane), aromatic (xylene, benzene, toluene) and petroleum (gasoline, diesel, kerosene, crude oil) compounds. It exhibited a maximum emulsification activity with weathered crude oil (97%) and was capable of transforming the rheological behavior of the pseudoplastic to a Newtonian fluid. The results reveal the potential of the bioemulsifier for use in bioremediation of hydrocarbons in marine environments and in enhanced oil recovery.
Sigma-Aldrich
Sodium benzoate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium benzoate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium benzoate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Supelco
Sodium benzoate, Pharmaceutical Secondary Standard; Certified Reference Material