MilliporeSigma
  • Dispersive liquid-liquid microextraction combined with microvolume spectrophotometry to turn green the 5530 APHA standard method for determining phenols in water and wastewater.
Dispersive liquid-liquid microextraction combined with microvolume spectrophotometry to turn green the 5530 APHA standard method for determining phenols in water and wastewater.
Talanta (2012-09-04)
Isela Lavilla, Sandra Gil, Marta Costas, Carlos Bendicho
ABSTRACT
In this work, a new method based on the combination of dispersive liquid-liquid microextraction (DLLME) with microvolume spectrophotometry has been developed as a greener and miniaturized alternative to the 5530 APHA standard method for determining phenols in water and wastewater. The method relies on the oxidative coupling of phenols with 4-aminoantipyrine (4-AAP). In order to preconcentrate the dye formed, the classical liquid-liquid extraction used in the 5530 APHA method (involving 500 mL of sample and 50 mL of trichloromethane) has been replaced by DLLME (with 5 mL of sample, 50 μL of trichloromethane and 200 μL of acetonitrile). After optimization, the method yielded limits of detection and quantification (0.8 and 2.5 μg L(-1), respectively) that were comparable with those obtained by the 5530 APHA standard method. Repeatability, expressed as relative standard deviation, was 5.2% (N=6), and the enrichment factor (EF) was 700. The proposed method was applied to the determination of phenols in different water samples and a wastewater with recoveries in the range 90-99%. The greenness profile was established in accordance with the suggestions made by the NEMI (National Environmental Methods Index). The absence of PBT (persistent bioaccumulative and toxic chemicals) and corrosive reagents and a drastic reduction of generated wastes can be emphasized.
Product Description
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%