MilliporeSigma
  • Orderly ultrathin films based on perylene/poly(N-vinyl carbazole) assembled with layered double hydroxide nanosheets: 2D fluorescence resonance energy transfer and reversible fluorescence response for volatile organic compounds.
Orderly ultrathin films based on perylene/poly(N-vinyl carbazole) assembled with layered double hydroxide nanosheets: 2D fluorescence resonance energy transfer and reversible fluorescence response for volatile organic compounds.
Advanced materials (Deerfield Beach, Fla.) (2012-09-01)
Zhen Li, Jun Lu, Shuangde Li, Shenghui Qin, Yumei Qin
ABSTRACT
Neutral poly(N-vinyl carbazole) (PVK) and perylene are coassembled within the interlayers of layered double hydroxide (LDH) nanosheets to form (perylene@PVK/LDH)(n) ultrathin films by the hydrogen-bond layer-by-layer assembly method. An efficient 2D fluorescence resonance energy transfer (FRET) process from PVK to perylene is demonstrated, and this FRET process can be inhibited/recovered reversibly by the adsorption/desorption of common volatile organic compounds (VOCs).
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mw ~1,100,000, powder
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mn 25,000-50,000