MilliporeSigma
  • Genome sequence of Ralstonia sp. strain PBA, a bacterium involved in the biodegradation of 4-aminobenzenesulfonate.
Genome sequence of Ralstonia sp. strain PBA, a bacterium involved in the biodegradation of 4-aminobenzenesulfonate.
Journal of bacteriology (2012-08-31)
Han Ming Gan, Teong Han Chew, Yea-Ling Tay, Siew Fen Lye, Adibah Yahya
ABSTRACT
Ralstonia sp. strain PBA was isolated from textile wastewater in a coculture with Hydrogenophaga sp. strain PBC. Here we present the assembly and annotation of its genome, which may provide further insights into the mechanism of its interaction with strain PBC during 4-aminobenzenesulfonate degradation.
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Millipore
Nitrate Reagent B, suitable for microbiology