MilliporeSigma
  • A new water···Na+ coordination motif in an unexpected diatrizoic acid disodium salt crystal form.
A new water···Na+ coordination motif in an unexpected diatrizoic acid disodium salt crystal form.
Chemical communications (Cambridge, England) (2012-08-31)
Katharina Fucke, Michael J G Peach, Judith A K Howard, Jonathan W Steed
ABSTRACT
The disodium salt of diatrizoic acid crystallises as a tetragonal channel hydrate structure. One of the incorporated water molecules is coordinated to three individual sodium cations in a unique geometry.
Sigma-Aldrich
Diatrizoic acid, Iodine-containing contrast agent