MilliporeSigma
  • [Nuclear-power-plant accidents: thyroid cancer incidence and radiation-related health effects from the Chernobyl accident].
[Nuclear-power-plant accidents: thyroid cancer incidence and radiation-related health effects from the Chernobyl accident].
Medecine sciences : M/S (2012-08-28)
Martin Schlumberger, Bernard Le Guen
ABSTRACT
Following the Chernobyl accident, enormous amounts of radioisotopes were released in the atmosphere and have contaminated surrounding populations in the absence of rapid protective countermeasures. The highest radiation doses were delivered to the thyroid gland, and the only direct consequence of radiation exposure observed among contaminated population is the increased incidence of thyroid cancers among subjects who were children in 1986 and who lived at that time in Belarus, Ukraine or Russia.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Gold etchant, standard