MilliporeSigma
  • Anti-inflammatory and immunomodulatory effects of iridoid glycosides from Paederia scandens (LOUR.) MERRILL (Rubiaceae) on uric acid nephropathy rats.
Anti-inflammatory and immunomodulatory effects of iridoid glycosides from Paederia scandens (LOUR.) MERRILL (Rubiaceae) on uric acid nephropathy rats.
Life sciences (2012-08-23)
Wenjing Zhu, Mingqun Pang, Liuyi Dong, Xueying Huang, Shuangmiao Wang, Lanlan Zhou
ABSTRACT
Uric acid nephropathy (UAN) is due to excessive uric acid, which leads to hyperuricemia and kidney damage via the deposition of urate microcrystals in the kidneys. Iridoid glycosides of Paederia scandens (IGPS) is a major active component isolated from the traditional Chinese herb P. scandens (LOUR.) MERRILL (Rubiaceae). This study aimed to evaluate the anti-inflammatory and immunomodulatory effects of IGPS and its mechanism on UAN rats. The experimental model of UAN rats was induced by using uricopoiesis promoter adenine and uricase inhibitor potassium oxonate (PO). Treatment groups received three different doses of IGPS, allopurinol (AP) and benzbromarone (BEN) daily for 24days respectively. The histopathology of renal tissues in UAN rats were assessed for conventional morphological evaluation. The nuclear factor-κBp65 (NF-κBp65), monocyte chemoattractant protein-1 (MCP-1) and α-smooth muscle actin (α-SMA) protein expression of renal tissues in UAN rats were investigated by immunohistochemistry. MCP-1 and α-SMA mRNA levels were monitored by method of reverse transcription polymerase chain reaction (RT-PCR). Treatment with IGPS significantly ameliorated UAN induced renal tissue injury, inhibited the biological activity of NF-κBp65, MCP-1 and α-SMA, and suppressed the mRNA expressions of MCP-1 and α-SMA. IGPS exerts a protective effect against renal injury in UAN rats, possesses anti-inflammatory and immunomodulatory effects by inactivating NF-κBp65 pathway transmembrane signal transduction, down regulating the expression of MCP-1 and α-SMA to modulate pro-inflammatory mediator production in nephropathy tissue to improve renal fibrosis in UAN rats.
Sigma-Aldrich
Asperuloside, ≥95% (LC/MS-ELSD)
Supelco
Benzbromarone, analytical standard
Benzbromarone, European Pharmacopoeia (EP) Reference Standard