MilliporeSigma
  • Small intestinal transplant mucosal necrosis associated with enteral sodium polystyrene sulfonate administration.
Small intestinal transplant mucosal necrosis associated with enteral sodium polystyrene sulfonate administration.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2012-08-21)
M Saginur, A Thiesen, A Lacson, D Bigam, J Yap
ABSTRACT
Diarrhea is a common manifestation of disease in recipients of intestinal transplants. Sodium Polystyrene Sulfonate administration has been associated with significant bowel injury. Recognizing the diagnosis requires clinical awareness and comprehensive review of intestinal biopsies. We present an illustrative case and discussion. It is the first reported case in a pediatric intestinal transplant patient with serial intestinal biopsies documenting the evolution of disease.
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~1,000,000, powder
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate), average Mw ~70,000
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~70,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~200,000, 30 wt. % in H2O
Sigma-Aldrich
Poly(sodium 4-styrenesulfonate) solution, average Mw ~1,000,000, 25 wt. % in H2O
Sigma-Aldrich
Poly(4-styrenesulfonic acid) solution, Mw ~75,000, 18 wt. % in H2O