MilliporeSigma
  • Syntheses, structures, and steady state and time resolved photophysical properties of a tetraiminodiphenol macrocyclic ligand and its dinuclear zinc(II)/cadmium(II) complexes with coordinating and noncoordinating anions.
Syntheses, structures, and steady state and time resolved photophysical properties of a tetraiminodiphenol macrocyclic ligand and its dinuclear zinc(II)/cadmium(II) complexes with coordinating and noncoordinating anions.
Inorganic chemistry (2012-08-09)
Samit Majumder, Leena Mandal, Sasankasekhar Mohanta
ABSTRACT
The work in the present investigation reports the syntheses, structures, steady state, and time-resolved photophysical properties of a tetraiminodiphenol macrocyclic ligand H(2)L and its eight dinuclear zinc(II) complexes and one cadmium(II) complex having composition [Zn(2)L(H(2)O)(2)](ClO(4))(2)·2CH(3)CN (1), [Zn(2)L(H(2)O)(2)](ClO(4))(2)·2dmf (2), [Zn(2)L(H(2)O)(2)](NO(3))(2)·2dmf (3), [Zn(2)LCl(2)] (4), [Zn(2)L(N(3))(2)] (5), [Zn(2)L(NCS)(2)] (6), [Zn(2)L(NCO)(2)] (7), [Zn(2)L(NCSe)(2)](2)·dmf (8), and [Cd(2)L(OAc)(2)] (9) with various coordinating and noncoordinating anions. The structures of all the complexes 1-9 have been determined by single-crystal X-ray diffraction. The noncovalent interactions in the complexes result in the generation of the following topologies: two-dimensional network in 1, 2, 4, 6, 7, 8, and 9; three-dimensional network in 5. Spectrophotometric and spectrofluorometric titrations of the diprotonated salt [H(4)L](ClO(4))(2) with triethylamine as well as with zinc(II) acetate and cadmium(II) acetate have been carried out, revealing fluorescence enhancement of the macrocyclic system by the base and the metal ions. Steady state fluorescence properties of [H(4)L](ClO(4))(2) and 1-9 have been studied and their quantum yields have been determined. Time resolved fluorescence behavior of [H(4)L](ClO(4))(2) and the dizinc(II) and dicadmium(II) complexes 1-9 have also been studied, and their lifetimes and radiative and nonradiative rate constants have been determined. The induced fluorescence enhancement of the macrocycle by zinc(II) and cadmium(II) is in line with the greater rate of increase of the radiative rate constants in comparison to the smaller rate of increase of nonradiative rate constants for the metal complexes. The fluorescence decay profiles of all the systems, being investigated here, that is, [H(4)L](ClO(4))(2) and 1-9, follow triexponential patterns, revealing that at least three conformers/components are responsible to exhibit the fluorescence decay behavior. The systems and studies in this report have been compared with those in the reports of the previously published similar systems, revealing some interesting aspects.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine hydrochloride, ≥99.0% (AT)