MilliporeSigma
Dettol poisoning and the need for airway intervention.
Hong Kong medical journal = Xianggang yi xue za zhi (2012-08-07)
P K Lam, C K Chan, M L Tse, F L Lau
ABSTRACT
OBJECTIVES. To (1) characterise the clinical features of Dettol poisoning on a territory-wide basis, (2) assess the need for airway intervention after such poisoning and its time frame after ingestion, and (3) identify predictors for such an intervention. DESIGN. Case series. SETTING. Sixteen accident and emergency departments in Hong Kong. PATIENTS. Patients with Dettol ingestion who presented within 48 hours of ingestion from July 2005 to June 2009, derived from the database of the Hong Kong Poison Information Centre. RESULTS. In all, 213 patient records were identified, of which 36 were excluded based on pre-defined criteria and 177 were analysed. Among the latter, the median age was 32 (range, 2-95) years and the male-to-female ratio was 1:2.7 (48:129). Intentional ingestion constituted the majority (95%) of cases. The most common symptoms were related to the local irritative/corrosive effects on the aero-digestive tract, such as gastro-intestinal upset and localised throat pain. Airway intervention was required in 14 (8%) patients. All interventions were performed within 12 hours of Dettol ingestion and three cases involved re-intubation after extubation. Univariate analysis showed that a Glasgow Coma Scale score of <8, older age, a larger amount ingested, lip swelling, lung crackles, and wheezing were all associated with airway intervention. In the multivariate analysis using forward stepwise logistic regression, only coma (Glasgow Coma Scale score of <8) remained statistically significant. CONCLUSIONS. Delayed airway obstruction (>12 hours after Dettol ingestion) is unlikely. For those who are intubated, careful assessment of airway adequacy before extubation is strongly recommended to avoid extubation failure and subsequent re-intubation. Patients in coma (Glasgow Coma Scale score of <8) should prompt airway intervention.
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, 99%
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, purum, ≥98.0% (T)