MilliporeSigma
  • In vivo genotoxicity of 1-methylnaphthalene from comprehensive toxicity studies with B6C3F1 gpt delta mice.
In vivo genotoxicity of 1-methylnaphthalene from comprehensive toxicity studies with B6C3F1 gpt delta mice.
The Journal of toxicological sciences (2012-08-07)
Meilan Jin, Aki Kijima, Yuta Suzuki, Daisuke Hibi, Yuji Ishii, Takehiko Nohmi, Akiyoshi Nishikawa, Kumiko Ogawa, Takashi Umemura
ABSTRACT
1-Methylnaphthalene (1-MN), a constituent of the polycyclic aromatic hydrocarbons (PAHs), is a lung carcinogen in mice. However, conventional genotoxicity tests such as the Ames test and sister chromatid exchange (SCE) test have yielded equivocal results. In the present study, the in vivo genotoxicity of 1-methylnaphthalene (1-MN) together with its toxicological profile was investigated in a 13-week repeated dose toxicity study of 1-MN using B6C3F1 gpt delta mice. In the serum biochemistry, significant increases in AST and ALP were observed in males of the 0.15% 1-MN group. From histopathological examination, the incidence of single cell necrosis in the liver was significantly increased in males of the 0.15% 1-MN group; however, no changes were observed in the lungs, the target organ of 1-MN. In an in vivo mutation assay, no changes in mutant frequencies of gpt and red/gam (Spi-) in lung DNA of 1-MN treated mice were observed at 13 weeks. In addition, there were no significant differences in the proliferating cell nuclear antigen (PCNA)-positive ratios in bronchiolar epithelial cells among the groups for either sex. These results suggest that 1-MN at a carcinogenic dose not induce overt toxicity for any organs and has no in vivo genotoxicity in the lungs.
Sigma-Aldrich
1-Methylnaphthalene, ≥95%
Sigma-Aldrich
1-Methylnaphthalene, 95%
Supelco
1-Methylnaphthalene, analytical standard