MilliporeSigma
  • Block copolymer assisted synthesis of porous α-Ni(OH)2 microflowers with high surface areas as electrochemical pseudocapacitor materials.
Block copolymer assisted synthesis of porous α-Ni(OH)2 microflowers with high surface areas as electrochemical pseudocapacitor materials.
Chemical communications (Cambridge, England) (2012-08-03)
Bishnu Prasad Bastakoti, Hou-Sheng Huang, Lin-Chi Chen, Kevin C-W Wu, Yusuke Yamauchi
ABSTRACT
Porous α-Ni(OH)(2) microflowers are successfully synthesized via a one-step aqueous-phase reaction assisted by block copolymers under mild conditions. The electrochemical measurement demonstrates that the α-Ni(OH)(2) microflowers calcined at 200 °C are capable to deliver a specific capacity of 1551 F g(-1) in 6 M KOH solution, suggesting their high potential as a novel electrochemical pseudocapacitor.
Sigma-Aldrich
Nickel(II) hydroxide