MilliporeSigma
Organocatalytic synthesis of quinine-functionalized poly(carbonate)s.
Biomacromolecules (2012-08-02)
Justin A Edward, Matthew K Kiesewetter, Hyunuk Kim, James C A Flanagan, James L Hedrick, Robert M Waymouth
ABSTRACT
The ring-opening polymerization of substituted cyclic carbonates with 1-(3,5-bis-trifluoromethyl-phenyl)-3-cyclohexyl-thiourea (TU)/1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) organocatalysts afford highly functionalized oligocarbonates. The fluorescent alkaloid quinine can be readily incorporated into the oligocarbonates either by initiation from quinine or by ring-opening polymerization of a quinine-functionalized cyclic carbonate (MTC-Q). Copolymerization of MTC-Q with a boc-protected guanidinium cyclic carbonate affords, after deprotection, highly water-soluble cationic copolymers functionalized with both quinine and pendant guanidinium groups. When multiple quinine groups are attached to the oligomers, they exhibit minimal fluorescence due to self-quenching. Upon hydrolysis, the fluorescence intensity increases, providing a potential strategy for monitoring the hydrolysis rates in real time.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard