MilliporeSigma
  • Biotechnological production of arbutins (α- and β-arbutins), skin-lightening agents, and their derivatives.
Biotechnological production of arbutins (α- and β-arbutins), skin-lightening agents, and their derivatives.
Applied microbiology and biotechnology (2012-07-31)
Dong-Ho Seo, Jong-Hyun Jung, Jae-Eun Lee, Eun-Jung Jeon, Wooki Kim, Cheon-Seok Park
ABSTRACT
Arbutins (α- and β-arbutins) are glycosylated hydroquinones that are commercially used in the cosmetic industry. These compounds have an inhibitory function against tyrosinase, a critical enzyme for generating pigments, which leads to the prevention of melanin formation, resulting in a whitening effect on the skin. Although β-arbutin is found in various plants including bearberry, wheat, and pear, α-arbutin and other arbutin derivatives are synthesized by chemical and enzymatic methods. This article presents a mini-review of recent studies on the production of α-arbutin and other α- and β-arbutin derivatives via enzymatic bioconversion methods. In addition, the structures of α- and β-arbutin derivatives and their biological activities are discussed. The catalytic characteristics of various enzymes used in the biosynthesis of arbutin derivatives are also reviewed.
Product Description
Arbutin, European Pharmacopoeia (EP) Reference Standard
Supelco
Arbutin, analytical standard
Sigma-Aldrich
Arbutin, ≥98% (HPLC)