MilliporeSigma
  • Sunitinib in refractory adrenocortical carcinoma: a phase II, single-arm, open-label trial.
Sunitinib in refractory adrenocortical carcinoma: a phase II, single-arm, open-label trial.
The Journal of clinical endocrinology and metabolism (2012-07-28)
Matthias Kroiss, Marcus Quinkler, Sarah Johanssen, Nielka P van Erp, Nienke Lankheet, Alexander Pöllinger, Katharina Laubner, Christian J Strasburger, Stefanie Hahner, Hans-Helge Müller, Bruno Allolio, Martin Fassnacht
ABSTRACT
Treatment of refractory adrenocortical carcinoma (ACC) is not established. Animal experiments pointed toward adrenal toxicity of sunitinib. The objective of the study was to determine the antitumor effects of sunitinib in refractory ACC. This was a phase II, open-label trial using a two-stage accrual design. The study was conducted at two tertiary referral centers. Thirty-eight patients with refractory ACC progressing after mitotane and one to three cytotoxic chemotherapies participated in the study. The intervention included sunitinib at a standard dose (50 mg/d, 4 wk on, 2 wk off). Response was defined as progression-free survival (PFS) of 12 wk or longer (first tumor evaluation). Thirty-five patients could be evaluated for response. Five patients experienced stable disease, 24 had progressive disease, and six patients died from ACC before the first evaluation (naïve estimate five of 35=14.3%, median unbiased response rate 15.4%, 95% confidence interval 5.0-33.4%). The median PFS was 2.8 months. In responders, PFS ranged between 5.6 and 11.2 months and overall survival between 14.0 and 35.5 months. Of 36 serious adverse events, only nine were possibly related to sunitinib. Concomitant mitotane appeared to negatively impact on outcome. Furthermore, a negative correlation between the serum concentrations of sunitinib plus its active metabolite N-desethylsunitinib (SU12662) and mitotane (r=-0.650; P=0.114) was observed in seven evaluable patients suggestive of a relevant drug interaction. Sunitinib has modest activity in advanced refractory ACC, which compares favorably with other targeted treatments in these patients. Sunitinib serum levels might have been profoundly reduced by mitotane induced cytochrome P450-3A4 activity attenuating its antitumor activity and adverse effects. Together these findings suggest that sunitinib deserves further investigation in mitotane-naïve ACC patients.
Product Description
Supelco
2,4′-DDD, PESTANAL®, analytical standard