MilliporeSigma
Facile synthesis of red-emitting lysozyme-stabilized Ag nanoclusters.
Nanoscale (2012-07-28)
Tingyao Zhou, Yunhe Huang, Wenbin Li, Zhimin Cai, Feng Luo, Chaoyong James Yang, Xi Chen
ABSTRACT
A facile approach was developed to prepare positively charged and red-emitting lysozyme-stabilized Ag nanoclusters (Lys-AgNCs) using NaBH₄ as a reducing agent at room temperature. The Lys-AgNCs can be applied in the highly selective detection of Hg²⁺.
Sigma-Aldrich
Borax Anhydrous, BioUltra, anhydrous, ≥99%
Sigma-Aldrich
Borax Anhydrous, puriss. p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis