MilliporeSigma
  • Population growth rate responses of Ceriodaphnia dubia to ternary mixtures of specific acting chemicals: pharmacological versus ecotoxicological modes of action.
Population growth rate responses of Ceriodaphnia dubia to ternary mixtures of specific acting chemicals: pharmacological versus ecotoxicological modes of action.
Environmental science & technology (2012-07-26)
Carlos Barata, María Fernández-San Juan, Maria Luisa Feo, Ethel Eljarrrat, Amadeu M V M Soares, Damià Barceló, Donald J Baird
ABSTRACT
When considering joint toxic apical effects at higher levels of biological organization, such as the growth of populations, the so-called pharmacological mode of action that relies on toxicological mechanistic effects on molecular target sites may not be relevant. Such effects on population growth rate will depend on the extent to which juvenile and adult survival rates and production rates (juvenile developmental rates and reproduction) are affected by toxic exposure and also by the sensitivity of population growth rates to life-history changes. In such cases, the ecotoxicological mode of action, defined as the crucial life-history trait processes and/or xenobiotic-life-history trait interactions underlying a toxicological effect on population growth rate, should be considered. Life-table response experiments with the crustacean Ceriodaphnia dubia exposed to single and ternary mixtures of nine compounds were conducted to test the hypothesis that joint effects on population growth rates could be predicted from the mixture constituent ecotoxicological mode of action. Joint effects of mixtures containing pharmacologically dissimilar compounds (cadmium, λ-cyhalothrin, and chlorpyrifos) that differentially affected life-history traits contributing to population growth rates were accurately predicted by the independent-action concept. Conversely, the concentration-addition concept accurately predicted joint effects of two different mixtures: one containing pharmacologically similar acting pyrethroids that also affected similarly life-history traits, the other one that included pharmacologically dissimilar compounds (3,4-dichloroaniline, sodium bromide, and fenoxycarb) acting mainly on reproduction rates. These results indicate that when assessing combined effects on population growth rate responses, selection of mixture toxicity conceptual models based on the ecotoxicological mode of action of mixture constituents provided more accurate predictions than those based on the pharmacological mode of action.
Product Description
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Fenoxycarb, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
3,4-Dichloroaniline, 98%
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%