MilliporeSigma
  • Hierarchical self-assembly of polycyclic heteroaromatic stars into snowflake patterns.
Hierarchical self-assembly of polycyclic heteroaromatic stars into snowflake patterns.
Angewandte Chemie (International ed. in English) (2012-07-24)
Stefan-S Jester, Eva Sigmund, Lisa M Röck, Sigurd Höger
ABSTRACT
Seeing stars: The two-dimensional patterns of the polycyclic heteroaromatic star molecules 1 on graphite vary with the side chain length. For n=12, frustrated self-assembly leads to hierarchically organized superstructures: up to 10 molecules form triangular aggregates which pack densely into hexagonal patterns with very large (15.5 nm) lattice constants.
Sigma-Aldrich
Triphenylene, 98%
Triphenylene, BCR®, certified reference material