MilliporeSigma
  • Evaluation of bicinchoninic acid as a ligand for copper(I)-catalyzed azide-alkyne bioconjugations.
Evaluation of bicinchoninic acid as a ligand for copper(I)-catalyzed azide-alkyne bioconjugations.
Organic & biomolecular chemistry (2012-07-24)
Erik H Christen, Raphael J Gübeli, Beate Kaufmann, Lars Merkel, Ronald Schoenmakers, Nediljko Budisa, Martin Fussenegger, Wilfried Weber, Birgit Wiltschi
ABSTRACT
The Cu(I)-catalyzed cycloaddition of terminal azides and alkynes (click chemistry) represents a highly specific reaction for the functionalization of biomolecules with chemical moieties such as dyes or polymer matrices. In this study we evaluate the use of bicinchoninic acid (BCA) as a ligand for Cu(I) under physiological reaction conditions. We demonstrate that the BCA-Cu(I)-complex represents an efficient catalyst for the conjugation of fluorophores or biotin to alkyne- or azide-functionalized proteins resulting in increased or at least equal reaction yields compared to commonly used catalysts like Cu(I) in complex with TBTA (tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine) or BPAA (bathophenanthroline disulfonic acid). The stabilization of Cu(I) with BCA represents a new strategy for achieving highly efficient bioconjugation reactions under physiological conditions in many application fields.
Sigma-Aldrich
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)