MilliporeSigma
  • Hispidin produced from Phellinus linteus protects against peroxynitrite-mediated DNA damage and hydroxyl radical generation.
Hispidin produced from Phellinus linteus protects against peroxynitrite-mediated DNA damage and hydroxyl radical generation.
Chemico-biological interactions (2012-07-24)
Wei Chen, Lina Feng, Zhaoyi Huang, Hongming Su
ABSTRACT
Oxidative stress plays an important role in the progression of many chronic diseases including cardiovascular diseases, diabetes, cancer and neurodegenerative disorders. One such mediator of oxidative stress is peroxynitrite, which is highly toxic to cultured neurons and astrocytes, and has been reported to be involved in the pathogenesis of various types of neuronal diseases. Therefore, searching for natural compounds with peroxynitrite-scavenging activity might be an effective therapy for peroxynitrite-mediated cytotoxicity. Hispidin, a phenolic compound from Phellinus linteus (a medicinal mushroom), has been shown to possess strong antioxidant, anticancer, and antidiabetic properties. However, the astrocyte protective efficacy of hispidin has been not examined. This study was undertaken to investigate whether the astrocyte protective effect of hispidin is associated with inhibition of peroxynitrite-induced DNA damage, a critical event leading to peroxynitrite-mediated cytotoxicity. Our results showed that peroxynitrite can cause DNA damage in φX-174 plasmid DNA and rat primary astrocytes. The presence of hispidin (10-20 μg/ml) was found to significantly inhibit peroxynitrite-induced DNA damage and cytotoxicity. EPR spectroscopy demonstrated that the formation of DMPO-hydroxyl radical adduct (DMPO-OH) from peroxynitrite, and that hispidin potently diminished the adduct signal in a concentration-dependent manner. Taken together, these results demonstrate for the first time that hispidin can protect against peroxynitrite-mediated cytotoxicity, DNA damage and hydroxyl radical formation.
Sigma-Aldrich
Hispidin, solid, ≥98% (HPLC)