MilliporeSigma
  • A sensitive microextraction by packed sorbent-based methodology combined with ultra-high pressure liquid chromatography as a powerful technique for analysis of biologically active flavonols in wines.
A sensitive microextraction by packed sorbent-based methodology combined with ultra-high pressure liquid chromatography as a powerful technique for analysis of biologically active flavonols in wines.
Analytica chimica acta (2012-07-24)
Catarina L Silva, João L Gonçalves, José S Câmara
ABSTRACT
A new approach based on microextraction by packed sorbent (MEPS) and reversed-phase high-throughput ultra high pressure liquid chromatography (UHPLC) method that uses a gradient elution and diode array detection to quantitate three biologically active flavonols in wines, myricetin, quercetin, and kaempferol, is described. In addition to performing routine experiments to establish the validity of the assay to internationally accepted criteria (selectivity, linearity, sensitivity, precision, accuracy), experiments are included to assess the effect of the important experimental parameters such as the type of sorbent material (C2, C8, C18, SIL, and C8/SCX), number of extraction cycles (extract-discard), elution volume, sample volume, and ethanol content, on the MEPS performance. The optimal conditions of MEPS extraction were obtained using C8 sorbent and small sample volumes (250μL) in five extraction cycle and in a short time period (about 5min for the entire sample preparation step). Under optimized conditions, excellent linearity (R(values)(2)>0.9963), limits of detection of 0.006μgmL(-1) (quercetin) to 0.013μgmL(-1) (myricetin) and precision within 0.5-3.1% were observed for the target flavonols. The average recoveries of myricetin, quercetin and kaempferol for real samples were 83.0-97.7% with relative standard deviation (RSD, %) lower than 1.6%. The results obtained showed that the most abundant flavonol in the analyzed samples was myricetin (5.8±3.7μgmL(-1)). Quercetin (0.97±0.41μgmL(-1)) and kaempferol (0.66±0.24μgmL(-1)) were found in a lower concentration. The optimized MEPS(C8) method was compared with a reverse-phase solid-phase extraction (SPE) procedure using as sorbent a macroporous copolymer made from a balanced ratio of two monomers, the lipophilic divinylbenzene and the hydrophilic N-vinylpyrrolidone (Oasis HLB) were used as reference. MEPS(C8) approach offers an attractive alternative for analysis of flavonols in wines, providing a number of advantages including highest extraction efficiency (from 85.9±0.9% to 92.1±0.5%) in the shortest extraction time with low solvent consumption, fast sample throughput, more environmentally friendly and easy to perform.
Product Description
Supelco
Myricetin, analytical standard
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)