MilliporeSigma
  • IBS-catalyzed regioselective oxidation of phenols to 1,2-quinones with Oxone®.
IBS-catalyzed regioselective oxidation of phenols to 1,2-quinones with Oxone®.
Molecules (Basel, Switzerland) (2012-07-20)
Muhammet Uyanik, Tatsuya Mutsuga, Kazuaki Ishihara
ABSTRACT
We have developed the first example of hypervalent iodine(V)-catalyzed regioselective oxidation of phenols to o-quinones. Various phenols could be oxidized to the corresponding o-quinones in good to excellent yields using catalytic amounts of sodium salts of 2-iodobenzenesulfonic acids (pre-IBSes) and stoichiometric amounts of Oxone® as a co-oxidant under mild conditions. The reaction rate of IBS-catalyzed oxidation under nonaqueous conditions was further accelerated in the presence of an inorganic base such as potassium carbonate (K₂CO₃), a phase transfer catalyst such as tetrabutylammonium hydrogen sulfate (nBu₄NHSO₄), and a dehydrating agent such as anhydrous sodium sulfate (Na₂SO₄).
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF