MilliporeSigma
Preparation and antibacterial activities of hollow silica-Ag spheres.
Colloids and surfaces. B, Biointerfaces (2012-07-17)
Lin Lin, Haifang Zhang, Haiying Cui, Mingqiang Xu, Shunsheng Cao, Guanghong Zheng, Mingdong Dong
ABSTRACT
Hollow silica spheres with round mesoporous shells were synthesized by core-shell template method, using monodispersed cationic polystyrene particles as core, and TEOS (tetraethoxysilane) as the silica source to form shell. After calcination at 550°C, uniform spheres with a thin shell of silica and hollow interior structures. Transmission electron microscopy results showed that the size of the spheres is about 700 nm in diameter with narrow distribution. In addition, the spheres have a high surface area of 183 m(2)/g. The spheres were subsequently used as silver-loading substrates and the silver loaded spheres were tested in antimicrobial study against gram negative bacteria Eschrichia coli in vitro. The hollow silica-Ag spheres proved significantly higher antibacterial efficacy against E. coli as compared to that of the common silica-Ag particles.
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems