MilliporeSigma
  • Constrained phenylalanine analogues. Preferred conformation of the 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) residue.
Constrained phenylalanine analogues. Preferred conformation of the 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid (Tic) residue.
International journal of peptide and protein research (1992-09-01)
G Valle, W M Kazmierski, M Crisma, G M Bonora, C Toniolo, V J Hruby
ABSTRACT
Three Tic-containing (Tic = 1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid) model peptides were synthesized to assess the tendency of this constrained Phe analogue to fold into a beta-bend and a helical structure, and to adopt a preferred side-chain disposition. The results of the solution conformational analysis, performed by using Fourier transform infrared absorption and 1H nuclear magnetic resonance, indicate that in chloroform the -Aib-D-Tic-Aib-, -(Aib)2-D-Tic-(Aib)2-, and -L-Pro-D-Tic- sequences fold into intramolecularly H-bonded forms to a great extent. An X-ray diffraction analysis on p-BrBz-(Aib)2-DL-Tic-(Aib)2-OMe monohydrate and p-BrBz-L-Pro-D-Tic-NHMe allows us to conclude that, while the pentapeptide methylester forms an incipient (distorted) 3(10)-helix, the dipeptide methylamide adopts a type-II beta-bend conformation. In both cases, the D-Tic side-chain conformation is D, gauche(-). The implications for the use of the Tic residue in designing conformationally restricted analogues of bioactive peptides are briefly discussed.
Sigma-Aldrich
(S)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, 97%
Sigma-Aldrich
1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid hydrochloride, 96%