MilliporeSigma
  • Cellular uptake and intracellular trafficking of PEG-b-PLA polymeric micelles.
Cellular uptake and intracellular trafficking of PEG-b-PLA polymeric micelles.
Biomaterials (2012-07-17)
Zhen Zhang, Xiaoqin Xiong, Jiangling Wan, Ling Xiao, Lu Gan, Youmei Feng, Huibi Xu, Xiangliang Yang
ABSTRACT
Besides as an inert carrier for hydrophobic anticancer agents, polymeric micelles composed of di-block copolymer poly(ethylene glycol)-poly(lactic acid) (PEG-b-PLA) function as biological response modifiers including reversal of multidrug resistance in cancer. However, the uptake mechanisms and the subsequent intracellular trafficking remain to be elucidated. In this paper, we found that the uptake of PEG-b-PLA polymeric micelles incorporating nile red (M-NR) was significantly inhibited by both dynamin inhibitor dynasore and dynamin-2 dominant negative mutant (dynamin-2 K44A). Exogenously expressed caveolin-1 colocalized with M-NR and upregulated M-NR internalization in HepG2 cells expressing low level of endogenous caveolin-1, while caveolin-1 dominant negative mutant (caveolin-1 Y14F) significantly downregulated M-NR internalization in C6 cells expressing high level of endogenous caveolin-1. Exogenously expressed clathrin light chain A (clathrin LCa) did not mainly colocalize with the internalized M-NR and had no effect on M-NR uptake. These results suggested that dynamin- and caveolin-dependent but clathrin-independent endocytosis was involved in M-NR cellular uptake. We further found that M-NR colocalized with lysosome and microtubulin after internalization.
Sigma-Aldrich
Nile Red, Technical grade
Sigma-Aldrich
Nile Red, for microscopy
Sigma-Aldrich
Nile Red, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)