MilliporeSigma
  • Spectinomycin resistance mutations in the rrn16 gene are new plastid markers in Medicago sativa.
Spectinomycin resistance mutations in the rrn16 gene are new plastid markers in Medicago sativa.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik (2012-07-14)
Brigitta Dudas, Barnabas Jenes, Gyorgy Botond Kiss, Pal Maliga
ABSTRACT
We report here the isolation of spectinomycin-resistant mutants in cultured cells of Medicago sativa line RegenSY-T2. Spectinomycin induces bleaching of cultured alfalfa cells due to inhibition of protein synthesis on the prokaryotic type 70S plastid ribosomes. Spontaneous mutants resistant to spectinomycin bleaching were identified by their ability to form green shoots on plant regeneration medium containing selective spectinomycin concentrations in the range of 25-50 mg/l. Sequencing of the plastid rrn16 gene revealed that spectinomycin resistance is due to mutations in a conserved stem structure of the 16S rRNA. Resistant plants transferred to the greenhouse developed normally and produced spectinomycin-resistant seed progeny. In light of their absence in soybean, a related leguminous plant, the isolation of spectinomycin-resistant mutants in M. sativa was unexpected. The new mutations are useful for the study of plastid inheritance, as demonstrated by detection of predominantly paternal plastid inheritance in the RegenSY-T2 × Szapko57 cross, and can be used as selective markers in plastid transformation vectors to obtain cisgenic plants.
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, γ-irradiated
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, powder, BioReagent, suitable for cell culture
Supelco
Spectinomycin dihydrochloride pentahydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Spectinomycin dihydrochloride pentahydrate, potency: ≥603 μg per mg