MilliporeSigma
  • Evaluation of cefoxitin disk diffusion breakpoint for detection of methicillin resistance in Staphylococcus pseudintermedius isolates from dogs.
Evaluation of cefoxitin disk diffusion breakpoint for detection of methicillin resistance in Staphylococcus pseudintermedius isolates from dogs.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (2012-07-13)
David A Bemis, Rebekah D Jones, Ricardo Videla, Stephen A Kania
ABSTRACT
Cefoxitin disk diffusion susceptibility testing is a recommended screening method for the detection of methicillin resistance in human isolates of Staphylococcus aureus and coagulase-negative staphylococci. A retrospective analysis of 1,146 clinical isolates of Staphylococcus pseudintermedius from dogs was conducted to determine if screening by the cefoxitin disk method can be similarly useful with S. pseudintermedius. The distribution of cefoxitin growth inhibition zone diameters within this collection was bimodal and correlated well with the results of methicillin resistance gene (mecA) detection by polymerase chain reaction. Of the isolates, 5% had discordant results and, when retested, 84% of these were in agreement. While a greater diversity of isolates and interlaboratory comparisons must be tested, the current study suggests that an epidemiological breakpoint (of approximately ≤ 30 mm = resistant; ≥ 31 = susceptible) can be established to predict methicillin resistance in S. pseudintermedius.
Sigma-Aldrich
Methicillin sodium salt, ≥95% (HPLC)
Supelco
Cefoxitin sodium salt
Cefoxitin sodium, European Pharmacopoeia (EP) Reference Standard