MilliporeSigma
  • Optimization of 4-aminoquinoline/clotrimazole-based hybrid antimalarials: further structure-activity relationships, in vivo studies, and preliminary toxicity profiling.
Optimization of 4-aminoquinoline/clotrimazole-based hybrid antimalarials: further structure-activity relationships, in vivo studies, and preliminary toxicity profiling.
Journal of medicinal chemistry (2012-07-13)
Sandra Gemma, Caterina Camodeca, Salvatore Sanna Coccone, Bhupendra P Joshi, Matteo Bernetti, Vittoria Moretti, Simone Brogi, Maria Cruz Bonache de Marcos, Luisa Savini, Donatella Taramelli, Nicoletta Basilico, Silvia Parapini, Matthias Rottmann, Reto Brun, Stefania Lamponi, Silvio Caccia, Giovanna Guiso, Robert L Summers, Rowena E Martin, Simona Saponara, Beatrice Gorelli, Ettore Novellino, Giuseppe Campiani, Stefania Butini
ABSTRACT
Despite recent progress in the fight against malaria, the emergence and spread of drug-resistant parasites remains a serious obstacle to the treatment of infections. We recently reported the development of a novel antimalarial drug that combines the 4-aminoquinoline pharmacophore of chloroquine with that of clotrimazole-based antimalarials. Here we describe the optimization of this class of hybrid drug through in-depth structure-activity relationship studies. Antiplasmodial properties and mode of action were characterized in vitro and in vivo, and interactions with the parasite's 'chloroquine resistance transporter' were investigated in a Xenopus laevis oocyte expression system. These tests indicated that piperazine derivatives 4b and 4d may be suitable for coadministration with chloroquine against chloroquine-resistant parasites. The potential for metabolism of the drugs by cytochrome P450 was determined in silico, and the lead compounds were tested for toxicity and mutagenicity. A preliminary pharmacokinetic analysis undertaken in mice indicated that compound 4b has an optimal half-life.
Product Description
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clotrimazole
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Clotrimazole, VETRANAL®, analytical standard
Clotrimazole, European Pharmacopoeia (EP) Reference Standard