MilliporeSigma
  • A comparison of indocyanine green fluorescence imaging plus blue dye and blue dye alone for sentinel node navigation surgery in breast cancer patients.
A comparison of indocyanine green fluorescence imaging plus blue dye and blue dye alone for sentinel node navigation surgery in breast cancer patients.
Annals of surgical oncology (2012-07-12)
Akira Hirano, Mari Kamimura, Kaoru Ogura, Naomi Kim, Akinori Hattori, Yumika Setoguchi, Fumie Okubo, Hiroaki Inoue, Reiko Miyamoto, Jun Kinoshita, Mariko Fujibayashi, Tadao Shimizu
ABSTRACT
To evaluate two methods of sentinel node navigation surgery (SNNS) using blue dye with and without indocyanine green (ICG) fluorescence imaging (FI) to determine the usefulness of combined ICG and blue dye. Between 2005 and 2010, a total of 501 patients underwent SNNS in our hospital. Detection of sentinel lymph node (SLN) was performed with sulfan blue (SB) alone until 2008 and with a combination of SB and ICG-FI since 2009. ICG 5 mg and SB 15 mg were injected in the subareolar region, and FI was obtained by a fluorescence imaging device. We attempted to identify SLNs in 393 patients by SB alone and in 108 patients by a combination of SB and FI. The mean number of SLNs detected was 1.6 (0-5) for SB alone and 2.2 (1-6) for the combination method. The SLN identification rate was 95.7 % for SB alone and 100 % for the combination method so that the combination was significantly superior to SB in terms of the identification rate (p = 0.0037). In patients who received the combination method, detection of SLN was made through only SB in 1 patient, only ICG in 8 patients, and both in 99 patients. Lymph node metastasis was found in 56 patients with SB alone and in 16 patients with the combination method. Recurrence of an axillary node was observed in 3 patients (0.8 %) with SB alone and in no patients with the combination method. ICG-FI is a useful method and is especially recommended in cases where no radiotracers are available.
Sigma-Aldrich
Patent blue VF sodium salt, for microscopy
Sigma-Aldrich
Patent Blue VF, Dye content 50 %