MilliporeSigma
  • Contribution of surfactin and SwrA to flagellin expression, swimming, and surface motility in Bacillus subtilis.
Contribution of surfactin and SwrA to flagellin expression, swimming, and surface motility in Bacillus subtilis.
Applied and environmental microbiology (2012-07-10)
Emilia Ghelardi, Sara Salvetti, Mara Ceragioli, Sokhna A Gueye, Francesco Celandroni, Sonia Senesi
ABSTRACT
Multicellular communities produced by Bacillus subtilis can adopt sliding or swarming to translocate over surfaces. While sliding is a flagellum-independent motility produced by the expansive forces in a growing colony, swarming requires flagellar functionality and is characterized by the appearance of hyperflagellated swarm cells that associate in bundles or rafts during movement. Previous work has shown that swarming by undomesticated B. subtilis strains requires swrA, a gene that upregulates the expression of flagellar genes and increases swimming motility, and surfactin, a lipopeptide biosurfactant that also facilitates sliding. Through an analysis of swrA(+) and swrA mutant laboratory strains with or without a mutation in sfp (a gene involved in surfactin production), we show that both swrA and surfactin upregulate the transcription of the flagellin gene and increase bacterial swimming. Surfactin also allows the nonswarming swrA mutant strain to efficiently colonize moist surfaces by sliding. Finally, we reconfirm the essential role of swrA in swarming and show that surfactin, which increases surface wettability, allows swrA(+) strains to produce swarm cells on media at low humidity.
Sigma-Aldrich
Surfactin, from Bacillus subtilis, ≥98.0% (HPLC)