MilliporeSigma
  • Determination of terbuthylazine and desethylterbuthylazine in human urine and hair samples by eletrospray ionization-liquid chromatography/triple quadrupole mass spectrometry.
Determination of terbuthylazine and desethylterbuthylazine in human urine and hair samples by eletrospray ionization-liquid chromatography/triple quadrupole mass spectrometry.
Analytical and bioanalytical chemistry (2012-07-04)
Rosa Mercadante, Elisa Polledri, Silvia Fustinoni
ABSTRACT
Terbuthylazine (TBA) is a widely applied herbicide and an environmental contaminant. Following its use, humans, such as agricultural workers and rural residents, may be exposed. An isotope-dilution liquid chromatography coupled to electrospray-tandem mass spectrometry method for the determination of TBA, and its metabolite desethylterbuthylazine (DET) in human urine and hair was developed and validated. Under the optimised conditions, analytes were extracted from urine using a solid phase cartridge or from hair by sonication in methanol. Analytes were separated using a C18 reversed-phase chromatographic column and quantified, after positive ionization using a heated electrospray source, by a triple quadrupole mass detector in the selected reaction monitoring mode. Validation showed linear dynamic ranges up to 100 μg/L or 5.00 ng/mg hair, inter- and intra-run precisions <7%, and accuracies within 12% of spiked concentrations. Limits of quantification were 0.25 μg/L in urine and 0.01 ng/mg hair for both TBA and DET. Matrix effect evaluation showed that the isotope dilution approach allowed for the control of bias sources. TBA and DET were determined in specimens of agriculture workers exposed to TBA using the validated method. Hair samples contained TBA levels in the low nanogram per milligram range, and urine samples contained DET levels in the low microgram per liter range. Conversely, TBA levels in urine samples and DET levels in hair samples were always below the limit of quantification.
Product Description
Supelco
Terbuthylazine, PESTANAL®, analytical standard
Pricing and availability is not currently available.