- Fluoxetine is a potent inhibitor of coxsackievirus replication.
Fluoxetine is a potent inhibitor of coxsackievirus replication.
Antimicrobial agents and chemotherapy (2012-07-04)
Jun Zuo, Kevin K Quinn, Steve Kye, Paige Cooper, Robert Damoiseaux, Paul Krogstad
PMID22751539
ABSTRACT
No antiviral drugs currently exist for the treatment of enterovirus infections, which are often severe and potentially life threatening. Molecular screening of small molecule libraries identified fluoxetine, a selective serotonin reuptake inhibitor, as a potent inhibitor of coxsackievirus replication. Fluoxetine did not interfere with either viral entry or translation of the viral genome. Instead, fluoxetine and its metabolite norfluoxetine markedly reduced the synthesis of viral RNA and protein. In view of its favorable pharmacokinetics and safety profile, fluoxetine warrants additional study as a potential antiviral agent for enterovirus infections.