MilliporeSigma
  • Synthesis and evaluation of uniformly sized nalidixic acid-imprinted nanospheres based on precipitation polymerization method for analytical and biomedical applications.
Synthesis and evaluation of uniformly sized nalidixic acid-imprinted nanospheres based on precipitation polymerization method for analytical and biomedical applications.
Journal of molecular recognition : JMR (2012-06-27)
Atefeh Abouzarzadeh, Mehdi Forouzani, Mohsen Jahanshahi, Nader Bahramifar
ABSTRACT
For the first time in this work, uniform molecularly imprinted polymer (MIP) nanoparticles were prepared using nalidixic acid as a template. The MIP nanoparticles were successfully synthesized by precipitation polymerization applying methacrylic acid (MAA) as a functional monomer and trimethylolpropane trimethacrylate (TRIM) as a cross-linking monomer at different mole ratios. The morphology, binding, recognition, selectivity, and in vitro release behaviors of obtained particles were studied. The produced polymers were characterized by Fourier transform infrared spectroscopy and differential scanning calorimetric. Furthermore, their morphology was analyzed accurately by scanning electron microscopy, photon correlation spectroscopy, and Brunauer-Emmett-Teller analysis. The nanospheres and microspheres with mean diameter values of 94 nm, 256 nm, and 1.2 µm were obtained using nalidixic acid-MAA-TRIM various mole ratios. Among the MIPs, the product with nalidixic acid-MAA-TRIM mole ratio of 1:12:12 established nanospheres with the lowest polydispersity index (0.003), an average pore diameter (12 nm), and the highest specific surface area (280 m(2) g(-1)) and selectivity factor (10.4). Results from binding experiments demonstrated that the imprinted nanospheres with a 94-nm mean diameter and a binding capacity of 28 mg of nalidixic acid per gram of polymer had higher specific affinity to nalidixic acid in contrast with the other imprinted nanospheres, microspheres, and nonimprinted particles. However, the binding performance of imprinted nanospheres in human serum was estimated using high-performance liquid chromatography analysis (binding approximately 98% of nalidixic acid). In addition, release experiments proved to be successful in the controlled release of nalidixic acid during a long period. The 20% of loaded nalidixic acid was released from the imprinted nanospheres within the first 20 h, whereas the remaining 80% was released in the after 120 h. The nalidixic acid release kinetics from the MIPs was highly affected by properties of the particles.
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Sigma-Aldrich
Nalidixic acid sodium salt, powder
Sigma-Aldrich
Nalidixic acid, ≥98%